Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ ACLY Pre-design Chimera RNAi
Homo sapiens ATP citrate lyase (ACLY), transcript variants, mRNA
Specifications
Content And Storage | Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity. |
---|---|
Description | Homo sapiens ATP citrate lyase (ACLY), transcript variants, mRNA. |
Species | Human |
For Use With (Application) | RNAi Knockdown |
Includes | DEPC water |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-035-706
|
Abnova™
H00000047R03S |
10 nmol | N/A | N/A | N/A | ||||
89-035-707
|
Abnova™
H00000047R03L |
20 nmol | N/A | N/A | N/A | ||||
Description
- Gene name: ACLY
- Gene Alias: ACL, ATPCL, CLATP
- Entrez GeneID: 47
- Gene Description: ATP citrate lyase
Specifications
Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity. | |
Human | |
DEPC water | |
Human |
Homo sapiens ATP citrate lyase (ACLY), transcript variants, mRNA. | |
RNAi Knockdown | |
ACLY Pre-design Chimera RNAi |
For research use only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title