Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AcmNPV gp64 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581761
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for AcmNPV Envelope glycoprotein gp64/AcmNPV-gp64.
Class III viral fusion protein. Envelope phosphoglycoprotein which mediates the fusion of viral and host endosomal membranes leading to virus entry into the host cell. After receptor-mediated internalization of the virus, gp64 undergoes conformational change into a fusion-competent state at low pH, and the nucleocapsid is released into the cytoplasm after cell fusion. May also play role in budding.
Specifications
AcmNPV gp64 | |
Polyclonal | |
Unconjugated | |
Gp64 | |
Recombinant AcmNPV Envelope glycoprotein gp64/AcmNPV-gp64 Protein. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction