Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AcmNPV gp64 Protein Antibody (06) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323537
Description
AcmNPV gp64 Protein Monoclonal antibody specifically detects AcmNPV gp64 Protein in Virus samples. It is validated for Western Blot, ELISASpecifications
AcmNPV gp64 Protein | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant AcmNPV gp64 Protein (Accession#: NP_054158.1; Met1-Thr481). | |
100 μL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA | |
06 | |
Western Blot 1:1000-1:5000, ELISA 1:1000-1:2000 | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction