Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AcmNPV gp64 Protein Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$140.50 - $423.50
Specifications
Antigen | AcmNPV gp64 Protein |
---|---|
Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
AcmNPV gp64 Protein Polyclonal antibody specifically detects AcmNPV gp64 Protein in Virus samples. It is validated for Western Blot, ELISASpecifications
AcmNPV gp64 Protein | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Virus | |
Rabbits immunized with purified, recombinant AcmNPV gp64 Protein (Accession#: NP_054158.1, Met1-Thr481) | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 | |
Polyclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
IgG | |
Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title