Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACMSD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582420
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582420 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582420 Supplier Invitrogen™ Supplier No. PA582420
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KGEAKLLKDG KVFRVVRENC WDPEVRIREM DQKGVTVQAL STVPVMFSYW AKPEDTLNLC QLLNNDLAST VVSYPRRFVG LGTLPMQAPE LAVKEMERCV KELGFPGVQI GTHVNEWDLN AQELFPVYAA AERLKCSLFV HPW Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 72%.

The neuronal excitotoxin quinolinate is an intermediate in the de novo synthesis pathway of NAD from tryptophan, and has been implicated in the pathogenesis of several neurodegenerative disorders. Quinolinate is derived from alpha-amino-beta-carboxy-muconate-epsilon-semialdehyde (ACMS). ACMSD (ACMS decarboxylase; EC 4.1.1.45) can divert ACMS to a benign catabolite and thus prevent the accumulation of quinolinate from ACMS.[supplied by OMIM, Oct 2004].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACMSD
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACMSD
Gene Accession No. Q8TDX5
Gene Alias 2-amino-3-carboxylmuconate-6-semialdehyde decarboxylase; 2-amino-3-carboxymuconate-6-semialdehyde decarboxylase; Acmsd; amino carboxymuconate semialdehyde decarboxylase; aminocarboxymuconate semialdehyde decarboxylase; Picolinate carboxylase; zgc:162888
Gene Symbols ACMSD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACMSD. Recombinant protein control fragment (Product #RP-90369).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130013
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.