Learn More
Invitrogen™ ACMSD Polyclonal Antibody
Description
Recommended positive controls: 293T, A431, H1299, HeLa, Molt-4, Raji, mouse brain. Predicted reactivity: Mouse (91%), Rat (89%), Zebrafish (87%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ACMSD |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.86 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | ACMSD |
| Gene Accession No. | Q8R519, Q8TDX5 |
| Gene Alias | 2-amino-3-carboxylmuconate-6-semialdehyde decarboxylase; 2-amino-3-carboxymuconate-6-semialdehyde decarboxylase; Acmsd; amino carboxymuconate semialdehyde decarboxylase; aminocarboxymuconate semialdehyde decarboxylase; Picolinate carboxylase; zgc:162888 |
| Gene Symbols | ACMSD |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.