Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACN9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554227
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554227 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554227 Supplier Invitrogen™ Supplier No. PA554227
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YATALLQQAN ENRQNSTGKA CFGTFLPEEK LNDFRDEQIG QLQELMQEAT KPNRQFSISE SMKP Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.

Plays an essential role in the assembly of succinate dehydrogenase (SDH), an enzyme complex (also referred to as respiratory complex II) that is a component of both the tricarboxylic acid (TCA) cycle and the mitochondrial electron transport chain, and which couples the oxidation of succinate to fumarate with the reduction of ubiquinone (coenzyme Q) to ubiquinol. Promotes maturation of the iron-sulfur protein subunit SDHB of the SDH catalytic dimer, protecting it from the deleterious effects of oxidants. May act together with SDHAF1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACN9
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SDHAF3
Gene Accession No. Q9NRP4
Gene Alias 0610005M07Rik; 4933430A16Rik; Acn9; ACN9 homolog; ACN9 homolog (S. cerevisiae); DC11; Liver regeneration-related protein LRRGT00092; LRRGT00092; LYRM10; protein ACN9 homolog, mitochondrial; SDH assembly factor 3; Sdh7; SDHAF3; Succinate dehydrogenase assembly factor 3, mitochondrial; succinate dehydrogenase complex assembly factor 3
Gene Symbols SDHAF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACN9. Recombinant protein control fragment (Product #RP-92844).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57001
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.