Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ acnB Monoclonal Antibody (09)
GREENER_CHOICE

Catalog No. PIMA528961
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528961 Supplier Invitrogen™ Supplier No. MA528961
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Escherichia coli acnB.

acnB is involved in the catabolism of short chain fatty acids (SCFA) via the tricarboxylic acid (TCA)(acetyl degradation route) and the 2-methylcitrate cycle I (propionate degradation route). It catalyzes the reversible isomerization of citrate to isocitrate via cis-aconitate. Also catalyzes the hydration of 2-methyl-cis-aconitate to yield (2R,3S)-2-methylisocitrate. The apo form of AcnB functions as a RNA-binding regulatory protein. During oxidative stress inactive AcnB apo-enzyme without iron sulfur clusters binds the acnB mRNA 3' UTRs (untranslated regions), stabilizes acnB mRNA and increases AcnB synthesis, thus mediating a post-transcriptional positive autoregulatory switch. AcnB also decreases the stability of the sodA transcript.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acnB
Applications ELISA
Classification Monoclonal
Clone 09
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias ACN; Aconitase
Host Species Mouse
Immunogen Recombinant Escherichia coli acnB.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.