Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aconitase 1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP151965
Description
Aconitase 1 Polyclonal specifically detects Aconitase 1 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
Aconitase 1 | |
Polyclonal | |
Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:8000 | |
NP_002188.1 | |
ACO1 | |
Peptide with sequence C-QVDFNRRADSLQKNQ corresponding to internal region according to NP_002188.1. | |
0.1 mg | |
metabolism | |
48 | |
Human | |
IgG |
Western Blot, ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
Aconitase, aconitase 1, soluble, aconitate hydratase, Citrate hydro-lyase, cytoplasmic aconitate hydratase, EC 4.2.1, EC 4.2.1.3, Ferritin repressor protein, IREB1IREBP1, IREBP, IRE-BP 1, Iron regulatory protein 1, iron-responsive element binding protein 1, Iron-responsive element-binding protein 1, IRP1ACONS | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog, and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction