Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Aconitase 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586346
Description
Purity is > 95% by SDS-PAGE.
Aconitase 1 (ACO1) also known as iron regulatory element binding protein 1 (IREB1), is a cytosolic protein which binds to iron-responsive elements (IREs). It plays a central role in cellular iron homeostasis. It was also shown to have aconitase activity, and hence grouped with the aconitase family of enzymes.Aconitase 1, also known as iron regulatory element binding protein 1 (IREB1), is a cytosolic protein which binds to iron-responsive elements (IREs). IREs are stem-loop structures found in the 5' UTR of ferritin mRNA, and in the 3' UTR of transferrin receptor mRNA. The iron-induced binding to the IRE results in repression of translation of ferritin mRNA, and inhibition of degradation of the otherwise rapidly degrading transferrin receptor mRNA. Thus, IREB1 plays a central role in cellular iron homeostasis. It was also shown to have aconitase activity, and hence grouped with the aconitase family of enzymes.
Specifications
Aconitase 1 | |
Polyclonal | |
Unconjugated | |
ACO1 | |
ACO1; Aco-1; Acon1; Aconitase; aconitase 1; aconitase 1, soluble; aconitate hydratase, cytoplasmic; ACONS; AH; Citrate hydro-lyase; cytoplasmic aconitase; cytoplasmic aconitate hydratase; cytosplasmic aconitase; epididymis luminal protein 60; ferritin repressor protein; FRP; HEL60; Ireb1; Irebp; IRE-BP 1; IREBP1; iron regulatory protein 1; iron-responsive element binding protein 1; iron-responsive element-binding protein 1; IRP1; OTTHUMP00000021176; OTTHUMP00000021177; OTTHUMP00000045233; soluble aconitase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11428, 48, 50655 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P21399, P28271, Q63270 | |
ACO1 | |
Synthetic peptide, corresponding to amino acids 680-720 of Human IRP-1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction