Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Aconitase 2 Monoclonal Antibody (7G4)
Mouse Monoclonal Antibody
$233.50 - $630.50
Specifications
Antigen | Aconitase 2 |
---|---|
Clone | 7G4 |
Concentration | 1 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Description
Aconitase 2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene belongs to the aconitase/IPM isomerase family. It is an enzyme that catalyzes the interconversion of citrate to isocitrate via cis-aconitate in the second step of the TCA cycle. This protein is encoded in the nucleus and functions in the mitochondrion. It was found to be one of the mitochondrial matrix proteins that are preferentially degraded by the serine protease 15(PRSS15), also known as Lon protease, after oxidative modification.Specifications
Aconitase 2 | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
Q99798, Q99KI0, Q9ER34 | |
11429, 50, 79250 | |
Full length human recombinant protein of human ACO2 (NP_001089) produced in HEK293T cell. | |
Antibody |
7G4 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Monoclonal | |
Liquid | |
IgG1 | |
Human, Mouse, Rat, Monkey | |
Aco2 | |
Aco2; Aco-2; Aco3; Aconitase; aconitase 2; aconitase 2, mitochondrial; aconitase precursor; aconitate hydratase, mitochondrial; ACONM; citrate hydro-lyase; D10Wsu183e; epididymis secretory sperm binding protein Li 284; heart aconitase; heart aconitase precursor; HEL-S-284; hypothetical protein; ICRD; MGC20605; MGC33908; mitochondrial aconitase; mitochondrial aconitase (nuclear aco2 gene); OCA8; OPA9; RP3-347H13.8 | |
Aco2 | |
Primary | |
Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title