Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aconitase 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586968
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586968 Supplier Invitrogen™ Supplier No. PA586968
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

The protein encoded by this gene belongs to the aconitase/IPM isomerase family. It is an enzyme that catalyzes the interconversion of citrate to isocitrate via cis-aconitate in the second step of the TCA cycle. This protein is encoded in the nucleus and functions in the mitochondrion. It was found to be one of the mitochondrial matrix proteins that are preferentially degraded by the serine protease 15(PRSS15), also known as Lon protease, after oxidative modification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aconitase 2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Aco2
Gene Accession No. Q99798, Q99KI0, Q9ER34
Gene Alias Aco2; Aco-2; Aco3; Aconitase; aconitase 2; aconitase 2, mitochondrial; aconitase precursor; aconitate hydratase, mitochondrial; ACONM; citrate hydro-lyase; D10Wsu183e; epididymis secretory sperm binding protein Li 284; heart aconitase; heart aconitase precursor; HEL-S-284; hypothetical protein; ICRD; MGC20605; MGC33908; mitochondrial aconitase; mitochondrial aconitase (nuclear aco2 gene); OCA8; OPA9; RP3-347H13.8
Gene Symbols Aco2
Host Species Rabbit
Immunogen Recombinant fusion protein of human Aconitase 2 (ACO2).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11429, 50, 79250
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.