Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aconitate hydratase B Antibody (02), DyLight 680 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306089FR
Description
Aconitate hydratase B Monoclonal antibody specifically detects Aconitate hydratase B in E. coli samples. It is validated for ELISA, ELISA, ELISASpecifications
Aconitate hydratase B | |
Monoclonal | |
DyLight 680 | |
acnB, hypothetical protein, yacI, yacJ | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Escherichia coli Aconitate hydratase B (P36683; Met1-Val865). | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
02 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
E. coli | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction