Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACOT1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | ACOT1 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB126587
|
Novus Biologicals
NBP310843100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
ACOT1 Polyclonal specifically detects ACOT1 in Mouse samples. It is validated for Western Blot.Specifications
ACOT1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
ACH2, acyl-CoA thioesterase 1Long chain acyl-CoA thioester hydrolase, acyl-coenzyme A thioesterase 1, CTE1, CTE-1, CTE-I, CTE-Ib, EC 3.1.2.2, Inducible cytosolic acyl-coenzyme A thioester hydrolase, LACH2, Long chain acyl-CoA hydrolase | |
The immunogen is a synthetic peptide directed towards the middle region of mouse ACOT1 (NP_599007.1). Peptide sequence KDIVDLLNNPLEGPDQKSLIPVERSDTAFLFLVGQDDHNWKSEFYAREAS | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
641371 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title