Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACOT11 Antibody (J4B2), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104289AF405
Description
ACOT11 Monoclonal specifically detects ACOT11 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ACOT11 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ACOT11 | |
Recombinant human ACOT11 (19-250aa) purified from E. coli | |
0.1 mL | |
Signal Transduction | |
26027 | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
J4B2 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Acyl-CoA thioester hydrolase 11, acyl-CoA thioesterase 11BFIT1, Adipose-associated thioesterase, BFIT2, BFITDKFZp667O1916, Brown fat-inducible thioesterase, EC 3.1.2.-, EC 3.1.2.1, KIAA0707acyl-coenzyme A thioesterase 11, STARD14, START domain containing 14, THEAStAR-related lipid transfer (START) domain containing 14, THEM1, thioesterase superfamily member 1, thioesterase, adipose associated | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human ACOT11 mAb, clone J4B2, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human ACOT11 protein. | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction