Learn More
Invitrogen™ ACOT11 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ACOT11 protein.
Specifications
Specifications
| Antigen | ACOT11 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ACOT11 |
| Gene Accession No. | Q8WXI4 |
| Gene Alias | 1110020M10Rik; 2010309H15Rik; ACOT11; Acyl-CoA thioester hydrolase 11; acyl-CoA thioesterase 11; Acyl-coenzyme A thioesterase 11; adipose-associated thioesterase; AW060409; Bfit; BFIT1; Brown fat-inducible thioesterase; KIAA0707; mKIAA0707; Palmitoyl-coenzyme A thioesterase; STARD14; StAR-related lipid transfer (START) domain containing 14; START domain containing 14; Thea; Them1; thioeste; thioesterase superfamily member 1; thioesterase, adipose associated |
| Gene Symbols | ACOT11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.