Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACOT11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103564
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103564 Supplier Invitrogen™ Supplier No. PA5103564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ACOT11.

This gene encodes a member of the acyl-CoA thioesterase family which catalyse the conversion of activated fatty acids to the corresponding non-esterified fatty acid and coenzyme A. Expression of a mouse homolog in brown adipose tissue is induced by low temperatures and repressed by warm temperatures. Higher levels of expression of the mouse homolog has been found in obesity-resistant mice compared with obesity-prone mice, suggesting a role of acyl-CoA thioesterase 11 in obesity. Alternative splicing results in transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACOT11
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACOT11
Gene Accession No. Q8VHQ9, Q8WXI4
Gene Alias 1110020M10Rik; 2010309H15Rik; ACOT11; Acyl-CoA thioester hydrolase 11; acyl-CoA thioesterase 11; Acyl-coenzyme A thioesterase 11; adipose-associated thioesterase; AW060409; Bfit; BFIT1; Brown fat-inducible thioesterase; KIAA0707; mKIAA0707; Palmitoyl-coenzyme A thioesterase; STARD14; StAR-related lipid transfer (START) domain containing 14; START domain containing 14; Thea; Them1; thioeste; thioesterase superfamily member 1; thioesterase, adipose associated
Gene Symbols ACOT11
Host Species Rabbit
Immunogen A synthesized peptide derived from human ACOT11(Accession Q8WXI4), corresponding to amino acid residues V249-G299.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100363074, 26027, 329910
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.