Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACOT12 Monoclonal Antibody (OTI1B9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525428
Description
ACOT12 Monoclonal Antibody for Western Blot, IHC (P), Flow
Hydrolyzes acetyl-CoA to acetate and CoA.
Specifications
ACOT12 | |
Monoclonal | |
0.91 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8WYK0 | |
ACOT12 | |
Full length human recombinant protein of ACOT12 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human, Monkey | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1B9 | |
Unconjugated | |
ACOT12 | |
1300004O04Rik; 4930449F15Rik; Acetyl-coenzyme A thioesterase; Acot12; Acyl-CoA thioester hydrolase 12; acyl-CoA thioesterase 12; Acyl-coenzyme A thioesterase 12; AV027244; Cach; CACH1; CACH-1; Cytoplasmic acetyl-CoA hydrolase 1; cytosolic acetyl-CoA hydrolase; hCACH-1; mCACH-1; rACH; rCACH-1; STARD15; StAR-related lipid transfer (START) domain containing 15; START domain-containing protein 15; THEAL | |
Mouse | |
Affinity Chromatography | |
RUO | |
134526 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction