Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACOT12 Monoclonal Antibody (OTI1D3), TrueMAB™, OriGene
SDP

Catalog No. 501673582
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-3582 100 μL
50-254-1481 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-3582 Supplier OriGene Supplier No. TA502530
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hydrolyzes acetyl-CoA to acetate and CoA.

Specifications

Antigen ACOT12
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1D3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ACOT12
Gene Accession No. Q8WYK0
Gene Alias CACH-1, Cach, STARD15, THEAL
Gene Symbols ACOT12
Host Species Mouse
Immunogen Full length human recombit protein of human ACOT12 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134526
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.