Learn More
Invitrogen™ ACOT2 Polyclonal Antibody

Description
Positive test controls include: 293T, LO2, U-87MG, Mouse kidney. The target is usually found in the following locations: Mitochondrion. Immunogen sequence: IVDMFGTGGG LLEYRASLLA GKGFAVMALA YYNYEDLPKT METLHLEYFE EAMNYLLSHP EVKGPGVGLL GISKGGELCL SMASFLKGIT AAVVINGSVA NVGGTLHYKG ETLPPVGVNR NRIKVTKDGY ADIVDVLNSP LEGPDQKSFI PVERAESTFL FLVGQDDHNW KSEFYANEAC KRLQAHGRRK PQIICYPETG HYIEPPYFPL CRASLHALVG SPIIWGGEPR AHAMAQVDAW KQLQTFFHKH LGGHEGTIPS KV.
Specifications
Specifications
| Antigen | ACOT2 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2.79 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | ACOT2 |
| Gene Accession No. | O55171, P49753, Q9QYR9 |
| Gene Alias | AA571646; Acot2; Acyl coenzyme A thioester hydrolase; acyl-CoA thioesterase 2; Acyl-coenzyme A thioester hydrolase 2a; acyl-coenzyme A thioesterase 2, mitochondrial; ARTISt/p43; CTE Ia; CTE1A; CTE-Ia; long-chain acyl-CoA thioesterase 2; mitochondrial acyl-CoA thioesterase 1; Mte1; MTE-I; peroxisomal long-chain acyl-coA thioesterase 2; PTE2; PTE2A; Very-long-chain acyl-CoA thioesterase; ZAP128 |
| Gene Symbols | ACOT2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.