Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACOT4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP183539
Description
ACOT4 Polyclonal specifically detects ACOT4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ACOT4 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Q8N9L9 | |
ACOT4 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:PPLGYDLRRIKVAFSGLVDIVDIRNALVGGYKNPSMIPIEKAQGPILLIVGQDDHNWRSELYAQTVSERLQAHGKEKPQIICYPGTGHYIEPPYFPLCPASLHRLLNKHVIWGGEPRAHSKAQEDAWKQILAFFCKHLGGTQKT | |
Affinity Purified | |
RUO | |
122970 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
acyl-CoA thioesterase 4EC 3.1.2.2, acyl-coenzyme A thioesterase 4, Peroxisomal acyl coenzyme A thioester hydrolase Ib, peroxisomal acyl-CoA thioesterase 2B, Peroxisomal long-chain acyl-CoA thioesterase Ib, PTE-2b, PTE2BPTE1B, PTEIB, PTE-IbFLJ31235 | |
Rabbit | |
46 kDa | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction