Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACOT4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549969
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549969 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549969 Supplier Invitrogen™ Supplier No. PA549969
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total ACOT4 protein.

Acyl-CoA thioesterases are a group of enzymes that catalyze the hydrolysis of acyl-CoAs to the free fatty acid and coenzyme A (CoASH), providing the potential to regulate intracellular levels of acyl-CoAs, free fatty acids and CoASH (PubMed:16940157). ACOT4 is a peroxisomal succinyl-coenzyme A thioesterase can also hydrolyze glutaryl-CoA and long chain saturated acyl-CoAs (PubMed:16940157). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACOT4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACOT4
Gene Accession No. Q8N9L9
Gene Alias ACOT4; Acyl-CoA thioesterase 4; acyl-coenzyme A thioesterase 4; B430212I04Rik; Peroxisomal acyl coenzyme A thioester hydrolase Ib; peroxisomal acyl-CoA thioesterase 2B; peroxisomal long-chain acyl-CoA thioesterase Ib; Peroxisomal succinyl-coenzyme A thioesterase; Pte1b; Pte2b; PTE-2b; PTEIB; PTE-Ib
Gene Symbols ACOT4
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human ACOT4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122970
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.