Learn More
Invitrogen™ ACOT4 Polyclonal Antibody

Description
Immunogen sequence: PPLGYDLRRI KVAFSGLVDI VDIRNALVGG YKNPSMIPIE KAQGPILLIV GQDDHNWRSE LYAQTVSERL QAHGKEKPQI ICYPGTGHYI EPPYFPLCPA SLHRLLNKHV IWGGEPRAHS KAQEDAWKQI LAFFCKHLGG TQKT Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 77%.
Specifications
Specifications
| Antigen | ACOT4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ACOT4 |
| Gene Accession No. | Q8N9L9 |
| Gene Alias | ACOT4; Acyl-CoA thioesterase 4; acyl-coenzyme A thioesterase 4; B430212I04Rik; Peroxisomal acyl coenzyme A thioester hydrolase Ib; peroxisomal acyl-CoA thioesterase 2B; peroxisomal long-chain acyl-CoA thioesterase Ib; Peroxisomal succinyl-coenzyme A thioesterase; Pte1b; Pte2b; PTE-2b; PTEIB; PTE-Ib |
| Gene Symbols | ACOT4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.