Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACOT6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570857
Description
This target displays homology in the following species: Cow: 86%; Horse: 93%; Human: 100%; Pig: 93%; Rat: 86%.
ACOT6 gene ontology annotations related to this gene include acyl-CoA metabolic process; fatty acid metabolic process.
Specifications
ACOT6 | |
Polyclonal | |
Unconjugated | |
Acot6 | |
4632408A20Rik; A330054B07; ACOT6; Acyl-CoA thioesterase 6; acyl-coenzyme A thioesterase 6; BE688602; c14_5530; C14orf42; Putative acyl-CoA thioesterase 6; putative acyl-coenzyme A thioesterase 6; RGD1309669 | |
Rabbit | |
Affinity chromatography | |
RUO | |
641372 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q3I5F7 | |
Acot6 | |
synthetic peptide directed towards the C-terminal region of human ACOT6. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction