Learn More
Invitrogen™ ACOT7 Monoclonal Antibody (1E7)

Description
Peptide Sequence: MSGPDVETPS AIQICRIMRP DDANVAGNVH GGTILKMIEE AGAIISTRHC NSQNGERCVA ALARVERTDF LSPMCIGEVA HVSAEITYTS KHSVEVQVNV MSENILTGAK KLTNKATLWY VPLSLKNVDK VLEVPPVVYS RQEQEEEGRK RYEAQKLERM ETKWRNGDIV QPVLNPEPNT VSYSQSSLIH LVGPSDCTLH GFVHGGVTMK LMDEVAGIVA ARHCKTNIVT ASVDAINFHD KIRKGCVITI SGRMTFTSNK SMEIEVLVDA DPVVDSSQKR YRAASAFFTY VSLSQEGRSL PVPQLVPETE DEKKRFEEGK GRYLQMKAKR QGHAEPQP.
Specifications
Specifications
| Antigen | ACOT7 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 1E7 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ACOT7 |
| Gene Accession No. | O00154 |
| Gene Alias | 2410041A17Rik; 50-kDa brain acyl-CoA hydrolase; ACH1; Acot7; ACT; acyl-CoA thioesterase 2; acyl-CoA thioesterase 7; acyl-CoA thioesterase, long chain; Bach; brain acyl CoA hydrolase; brain acyl-CoA hydrolase; CTE-II; CTE-IIa; CTE-IIb; cytosolic acyl coenzyme A thioester hydrolase; hBACH; L RP1-120G22.10; LACH; LACH1; Long chain acyl-CoA thioester hydrolase; MTE-II |
| Gene Symbols | ACOT7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.