Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACOT7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555251
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555251 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555251 Supplier Invitrogen™ Supplier No. PA555251
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLCEFGRQAS SRRLVAGQGC VGPRRGCCAP VQVVGPRADL PPCGACITGR IMRPDDANVA GNVHGGTILK MIEEAGAIIS TRHCNSQNGE Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 65%.

This gene encodes a member of the acyl coenzyme family. The encoded protein hydrolyzes the CoA thioester of palmitoyl-CoA and other long-chain fatty acids. Decreased expression of this gene may be associated with mesial temporal lobe epilepsy. Alternatively spliced transcript variants encoding distinct isoforms with different subcellular locations have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACOT7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACOT7
Gene Accession No. O00154
Gene Alias 2410041A17Rik; 50-kDa brain acyl-CoA hydrolase; ACH1; Acot7; ACT; acyl-CoA thioesterase 2; acyl-CoA thioesterase 7; acyl-CoA thioesterase, long chain; Bach; brain acyl CoA hydrolase; brain acyl-CoA hydrolase; CTE-II; CTE-IIa; CTE-IIb; cytosolic acyl coenzyme A thioester hydrolase; hBACH; L RP1-120G22.10; LACH; LACH1; Long chain acyl-CoA thioester hydrolase; MTE-II
Gene Symbols ACOT7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACOT7. Recombinant protein control fragment (Product #RP-94055).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11332
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.