Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACOT8 Antibody (4D10), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259419PE
This item is not returnable.
View return policy
Description
ACOT8 Monoclonal antibody specifically detects ACOT8 in Human samples. It is validated for Flow CytometrySpecifications
ACOT8 | |
Monoclonal | |
PE | |
ACTEIII, acyl-CoA thioesterase 8HNAACTE, Choloyl-coenzyme A thioesterase, EC 3.1.2.27, hACTEIII, HIV-Nef-associated acyl-CoA thioesterase, hTEcholoyl-CoA hydrolase, long-chain fatty-acyl-CoA hydrolase, palmitoyl-CoA hydrolase, peroxisomal acyl-CoA thioesterase, peroxisomal acyl-CoA thioesterase 1, Peroxisomal acyl-coenzyme A thioester hydrolase 1, Peroxisomal long-chain acyl-CoA thioesterase 1, PTE-1, PTE1HIV-Nef associated acyl-CoA thioesterase, PTE2, PTE-2hACTE-IIIacyl-coenzyme A thioesterase 8, Thioesterase II, thioesterase III | |
Recombinant human ACOT8 (1-319aa) purified from E. coli | |
0.1 mL | |
Cancer, Cardiovascular Biology, Endocrinology, Immunology, Signal Transduction | |
10005 | |
Store at 4°C in the dark. | |
IgG2a κ |
Flow Cytometry | |
4D10 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction