Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACOT8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526152
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526152 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526152 Supplier Invitrogen™ Supplier No. PA526152
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACOT8 Polyclonal Antibody for Western Blot, IHC (P), Flow

Acyl-CoA thioesterases are a group of enzymes that catalyze the hydrolysis of acyl-CoAs to the free fatty acid and coenzyme A (CoASH), providing the potential to regulate intracellular levels of acyl-CoAs, free fatty acids and CoASH. It may mediate Nef-induced down-regulation of CD4. It may be involved in the metabolic regulation of peroxisome proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACOT8
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ACOT8
Gene Accession No. O14734, P58137
Gene Alias 4,8-dimethylnonanoyl-CoA thioesterase; 48-dimethylnonanoyl-CoA thioesterase; ACOT8; ACTEIII; Acyl-CoA thioesterase 8; acyl-coenzyme A thioesterase 8; choloyl-CoA hydrolase; choloyl-coenzyme A thioesterase; hACTE-III; HIV-Nef associated acyl-CoA thioesterase; HIV-Nef-associated acyl-CoA thioesterase; HNAACTE; hTE; long-chain fatty-acyl-CoA hydrolase; NAP1; Nef (lentivirus myristoylated factor) associated protein 1; palmitoyl-CoA hydrolase; peroxisomal acyl-CoA thioesterase 1; peroxisomal acyl-CoA thioesterase 2; Peroxisomal acyl-coenzyme A thioester hydrolase 1; peroxisomal long-chain acyl-CoA thioesterase 1; PTE1; PTE-1; PTE2; PTE-2; Thioesterase II; thioesterase III
Gene Symbols ACOT8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 290-319 amino acids from the C-terminal region of human ACOT8.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10005, 170789
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.