Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACOT8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588959
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588959 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588959 Supplier Invitrogen™ Supplier No. PA588959
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSSPQAPEDG QGCGDRGDPP GDLRSVLVTT VLNLEPLDED LFRGRHYWVP AKRLFGGQIV GQALVAAAKS VSEDVHVHSL HCYFVRAGDP KLPVLYQVER TRTGSSFSVR SVKAVQHGKP IFICQASFQQ AQPSPMQHQF SMPTVPPPEE LLDCETLIDQ YLRDPNLQKR YPLALNRIAA QEVPIEIKPV NPSPLSQLQR MEPKQMFWVR ARGYIGEGDM; Positive Samples: Jurkat, Raji, U-87MG, HeLa; Cellular Location: Peroxisome.

Acyl-CoA thioesterases are a group of enzymes that catalyze the hydrolysis of acyl-CoAs to the free fatty acid and coenzyme A (CoASH), providing the potential to regulate intracellular levels of acyl-CoAs, free fatty acids and CoASH. It may mediate Nef-induced down-regulation of CD4. It may be involved in the metabolic regulation of peroxisome proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACOT8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ACOT8
Gene Accession No. O14734
Gene Alias 4,8-dimethylnonanoyl-CoA thioesterase; 48-dimethylnonanoyl-CoA thioesterase; ACOT8; ACTEIII; Acyl-CoA thioesterase 8; acyl-coenzyme A thioesterase 8; choloyl-CoA hydrolase; choloyl-coenzyme A thioesterase; hACTE-III; HIV-Nef associated acyl-CoA thioesterase; HIV-Nef-associated acyl-CoA thioesterase; HNAACTE; hTE; long-chain fatty-acyl-CoA hydrolase; NAP1; Nef (lentivirus myristoylated factor) associated protein 1; palmitoyl-CoA hydrolase; peroxisomal acyl-CoA thioesterase 1; peroxisomal acyl-CoA thioesterase 2; Peroxisomal acyl-coenzyme A thioester hydrolase 1; peroxisomal long-chain acyl-CoA thioesterase 1; PTE1; PTE-1; PTE2; PTE-2; Thioesterase II; thioesterase III
Gene Symbols ACOT8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ACOT8 (NP_0054602).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10005
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.