Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACOX1 Antibody (004), DyLight 680 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289865FR
Description
ACOX1 Monoclonal antibody specifically detects ACOX1 in Human samples. It is validated for ELISASpecifications
ACOX1 | |
Monoclonal | |
DyLight 680 | |
ACOXEC 1.3.3.6, acyl-CoA oxidase 1, palmitoyl, acyl-CoA oxidase, straight-chain, acyl-Coenzyme A oxidase 1, palmitoyl, AOX, PALMCOX, Palmitoyl-CoA oxidase, peroxisomal acyl-coenzyme A oxidase 1, peroxisomal fatty acyl-CoA oxidase, SCOXMGC1198, Straight-chain acyl-CoA oxidase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ACOX1 (AAH08767.1; Met1-Leu660). | |
0.1 mL | |
Lipid and Metabolism | |
51 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
004 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction