Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACOX1 Mouse anti-Human, Clone: 153CT43.1.1, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524715
Description
The protein encoded by this gene is the first enzyme of the fatty acid beta-oxidation pathway, which catalyzes the desaturation of acyl-CoAs to 2-trans-enoyl-CoAs. It donates electrons directly to molecular oxygen, thereby producing hydrogen peroxide. Defects in this gene result in pseudoneonatal adrenoleukodystrophy, a disease that is characterized by accumulation of very long chain fatty acids. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
ACOX1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q15067 | |
ACOX1 | |
human ACOX1 recombit protein | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
153CT43.1.1 | |
Unconjugated | |
ACOX1 | |
Peroxisomal acyl-coenzyme A oxidase 1, AOX, Palmitoyl-CoA oxidase, Straight-chain acyl-CoA oxidase, SCOX, ACOX1, ACOX | |
Mouse | |
Protein G | |
RUO | |
51 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction