Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACOX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584673
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584673 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584673 Supplier Invitrogen™ Supplier No. PA584673
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTVKGFTEAL EKLENEPAIQ QVLKRLCDLH AIHGILTNSG DFLHDAFLSG AQVDMARTAY LDLLRLIRKD AILLTDAFDF TDQCL Highest antigen sequence indentity to the following orthologs: Mouse - 79%, Rat - 79%.

The product of this gene belongs to the acyl-CoA oxidase family. It encodes the branched-chain acyl-CoA oxidase which is involved in the degradation of long branched fatty acids and bile acid intermediates in peroxisomes. Deficiency of this enzyme results in the accumulation of branched fatty acids and bile acid intermediates, and may lead to Zellweger syndrome, severe mental retardation, and death in children.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACOX2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACOX2
Gene Accession No. Q99424
Gene Alias 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholestanoyl-CoA 24-hydroxylase; 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholestanoyl-CoA oxidase; Acox2; acyl-CoA oxidase 2; acyl-CoA oxidase 2, branched chain; acyl-Coenzyme A oxidase 2, branched chain; BCOX; branched chain acyl-CoA oxidase; BRCACOX; BRCOX; Peroxisomal acyl-coenzyme A oxidase 2; peroxisomal branched chain acyl-CoA oxidase; THCA-CoA oxidase; THCCox; trihydroxycoprostanoyl-CoA oxidase
Gene Symbols ACOX2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACOX2. Recombinant protein control fragment (Product #RP-105345).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8309
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.