Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACOX3 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298600MFV500
Description
ACOX3 Polyclonal antibody specifically detects ACOX3 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
ACOX3 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
acyl-CoA oxidase 3, pristanoyl, Branched-chain acyl-CoA oxidase, BRCACox, BRCOX, EC 1.3.3.6, peroxisomal acyl-coenzyme A oxidase 3, PRCOX, pristanoyl, Pristanoyl-CoA oxidase | |
Produced in rabbits immunized with E. coli-derived Human ACOX3 fragment. | |
0.1 mL | |
Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
8310 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction