Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACOX3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | ACOX3 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ACOX3 Polyclonal specifically detects ACOX3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ACOX3 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
acyl-CoA oxidase 3, pristanoyl, Branched-chain acyl-CoA oxidase, BRCACox, BRCOX, EC 1.3.3.6, peroxisomal acyl-coenzyme A oxidase 3, PRCOX, pristanoyl, Pristanoyl-CoA oxidase | |
ACOX3 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Human | |
O15254 | |
8310 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ILNLKLAVAIALRFSATRRQFGPTEEEEIPVLEYPMQQWRLLPYLAAVYALDHFSKSLFLDLVELQRGLASGDRSARQAELGREIHALASASKP | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
78 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title