Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACOX3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557422
Description
Immunogen sequence: YRGGYFSGEQ AGEVLESAVL ALCSQLKDDA VALVDVIAPP DFVLDSPIGR ADGELYKNLW GAVLQESKVL ERASWWPEFS VNKPVIGSLK SKL Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 75%.
Acyl-Coenzyme A oxidase 3 also know as pristanoyl-CoA oxidase (ACOX3) is involved in the desaturation of 2-methyl branched fatty acids in peroxisomes. Unlike the rat homolog, the human gene is expressed in very low amounts in liver such that its mRNA was undetectable by routine Northern-blot analysis or its product by immunoblotting or by enzyme activity measurements. However the human cDNA encoding a 700 amino acid protein with a peroxisomal targeting C-terminal tripeptide S-K-L was isolated and is thought to be expressed under special conditions such as specific developmental stages or in a tissue specific manner in tissues that have not yet been examined.
Specifications
ACOX3 | |
Polyclonal | |
Unconjugated | |
ACOX3 | |
Acox3; acyl-CoA oxidase 3, pristanoyl; acyl-Coenzyme A oxidase 3, pristanoyl; BI685180; branched-chain acyl-CoA oxidase; BRCACox; BRCOX; EST-s59; PCOX; Peroxisomal acyl-coenzyme A oxidase 3; PRCOX; Pristanoyl-CoA oxidase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8310 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15254 | |
ACOX3 | |
Recombinant protein corresponding to Human ACOX3. Recombinant protein control fragment (Product #RP-96611). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction