Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACOX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112305
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112305 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112305 Supplier Invitrogen™ Supplier No. PA5112305
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACOX3 Polyclonal Antibody for Western Blot, IP

Acyl-Coenzyme A oxidase 3 also know as pristanoyl-CoA oxidase (ACOX3) is involved in the desaturation of 2-methyl branched fatty acids in peroxisomes. Unlike the rat homolog, the human gene is expressed in very low amounts in liver such that its mRNA was undetectable by routine Northern-blot analysis or its product by immunoblotting or by enzyme activity measurements. However the human cDNA encoding a 700 amino acid protein with a peroxisomal targeting C-terminal tripeptide S-K-L was isolated and is thought to be expressed under special conditions such as specific developmental stages or in a tissue specific manner in tissues that have not yet been examined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACOX3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene ACOX3
Gene Accession No. O15254
Gene Alias Acox3; acyl-CoA oxidase 3, pristanoyl; acyl-Coenzyme A oxidase 3, pristanoyl; BI685180; branched-chain acyl-CoA oxidase; BRCACox; BRCOX; EST-s59; PCOX; Peroxisomal acyl-coenzyme A oxidase 3; PRCOX; Pristanoyl-CoA oxidase
Gene Symbols ACOX3
Host Species Rabbit
Immunogen E. coli-derived Human ACOX3 fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8310
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.