Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACOXL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570858
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570858 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570858 Supplier Invitrogen™ Supplier No. PA570858
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 85%; Horse: 91%; Human: 100%; Pig: 83%; Rabbit: 92%.

ACOXL gene ontology annotations related to this gene include fatty acid beta-oxidation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACOXL
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ACOXL
Gene Accession No. Q9NUZ1
Gene Alias ACOXL; acyl-CoA oxidase like; acyl-CoA oxidase-like; acyl-CoA oxidase-like protein; acyl-Coenzyme A oxidase-like; acyl-coenzyme A oxidase-like protein; acyl-coenzyme A oxidase-like protein-like
Gene Symbols ACOXL
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human ACOXL.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55289
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.