Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACP5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987263
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7263 Supplier Bioss Supplier No. BS6434R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher′s disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin′s lymphoma and in the sera of patients undergoing active bone turnover.

Specifications

Antigen ACP5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ACP5
Gene Accession No. P13686
Gene Alias acid phosphatase 5, tartrate resistant; acid phosphatase type V; Acp5; HPAP; human purple acid phosphatase; T5ap; Tartrate-resistant acid ATPase; tartrate-resistant acid phosphatase 5a; tartrate-resistant acid phosphatase 5b; Tartrate-resistant acid phosphatase type 5; TRACP; TRACP5a; TRACP5b; Trap; TR-AP; TRAPase; TrATPase; TTRRAP; Type 5 acid phosphatase
Gene Symbols ACP5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Tartrate-resistant acid phosphatase type 5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.