Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACP6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | ACP6 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ACP6 Polyclonal specifically detects ACP6 in Human samples. It is validated for Western Blot.Specifications
| ACP6 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| acid phosphatase 6, lysophosphatidicPACPL1, Acid phosphatase-like protein 1, ACPL1acid phosphatase like 1, EC 3.1.3.2, LPAPlysophosphatidic acid phosphatase type 6 | |
| ACP6 | |
| IgG | |
| 49 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9NPH0 | |
| 51205 | |
| Synthetic peptides corresponding to ACP6(acid phosphatase 6, lysophosphatidic) The peptide sequence was selected from the N terminal of ACP6. Peptide sequence EADGQCPVDRSLLKLKMVQVVFRHGARSPLKPLPLEEQVEWNPQLLEVPP. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title