Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACPL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553567
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553567 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553567 Supplier Invitrogen™ Supplier No. PA553567
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STPKNGMSSK SRKRIMPDPV TEPPVTDPVY EALLYCNIPS VAERSMEGHA PHHFKLVSVH VFIRHGDRYP LYVIPKTKRP EIDCTLVANR KPYHPKLEAF ISHMSKGSGA SFESPLNSLP LYPNHP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.

Responsible for the 2-O-dephosphorylation of xylose in the glycosaminoglycan-protein linkage region of proteoglycans thereby regulating the amount of mature glycosaminoglycan (GAG) chains. Sulfated glycosaminoglycans (GAGs), including heparan sulfate and chondroitin sulfate, are synthesized on the so-called common GAG-protein linkage region (GlcUAbeta1-3Galbeta1-3Galbeta1-4Xylbeta1-O-Ser) of core proteins, which is formed by the stepwise addition of monosaccharide residues by the respective specific glycosyltransferases. Xylose 2-O-dephosphorylation during completion of linkage region formation is a prerequisite for the initiation and efficient elongation of the repeating disaccharide region of GAG chains. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACPL2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PXYLP1
Gene Accession No. Q8TE99
Gene Alias 2-phosphoxylose phosphatase 1; 9430094M07Rik; acid phosphatase-like 2; acid phosphatase-like protein 2; ACPL2; BB177120; C130099A20Rik; epididymis luminal protein 124; HEL124; PXYLP1; PXYLP1 (HGNC:26303); UNQ370/PRO706; Xylosyl phosphatase; XYLP
Gene Symbols PXYLP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACPL2. Recombinant protein control fragment (Product #RP-90339).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92370
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.