Learn More
Waters Corporation ACQUITY BEH130 C18 Method Validation Kit
Shop All Waters Chromatography Solutions Products
Description
- The small-pore (130 Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
Specifications
Specifications
| Diameter (Metric) | 2.1 mm |
| Length (Metric) | 100 mm |
| Particle Shape | Spherical |
| Particle Size | 1.7 μm |
| Pore Size | 130 Å |
| Quantity | 3/Pk. |
| USP Type | L1 |
| Endcapped | Yes |
| For Use With (Application) | Peptides |
| Product Type | Method Validation Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.