Learn More
Waters Corporation ACQUITY CSH C18 Method Transfer Kit
Catalog No. 50791492 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Particle Size:
1.7 to 3.5 μm
1.7 to 5 μm
Product Type:
Method Transfer Kit
Method Transfer Kit, High Res
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size Product Type
50-791-492 1.7 to 3.5 μm Method Transfer Kit, High Res
50-791-489 1.7 to 3.5 μm Method Transfer Kit
50-791-486 1.7 to 5 μm Method Transfer Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-791-492 Supplier Waters Corporation Supplier No. 186005535
ACQUITY Method Transfer Kits contain an ACQUITY CSH column and matching XSelect HPLC.
  • The columns contained in an ACQUITY CSH Method Transfer Kit possess the same length to particle size (L/dp) ratio and will provide the same selectivity.
Specifications
Particle Shape Spherical
Particle Size 1.7 to 3.5 μm
Pore Size 130 Å
Quantity 2/Pk.
USP Type L1
Endcapped Yes
Product Type Method Transfer Kit, High Res