Learn More
Waters Corporation ACQUITY CSH Fluoro-Phenyl Method Transfer Kit
Shop All Waters Chromatography Solutions Products
Description
- The columns contained in an ACQUITY CSH Method Transfer Kit possess the same length to particle size (L/dp) ratio and will provide the same selectivity.
Specifications
Specifications
| Particle Shape | Spherical |
| Particle Size | 1.7 to 3.5 μm |
| Pore Size | 130 Å |
| Pore Volume | 2/Pk. |
| Quantity | 2/Pk. |
| USP Type | L43 |
| Endcapped | No |
| Product Type | Method Transfer Kit, High Res |