Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corporation ACQUITY UPLC BEH200 SEC Column

Catalog No. NC0056764 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
300 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
NC0056764 300 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC0056764 Supplier Waters Corporation Supplier No. 186005226
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ACQUITY UPLC BEH200 SEC column based upon the highly successful BEH particle technology.

  • Fully optimized for the analysis of proteins and their aggregates with molecular weights ranging from 10,000 to 450,000 Daltons, the total system solution will routinely deliver fast, accurate aggregate determinations up to 10 times quicker that traditional HPLC-based SEC methods.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 300 mm
Particle Shape Spherical
Particle Size 1.7 μm
Pore Size 200 Å
Quantity 1/Ea.
USP Type L33
Endcapped No
For Use With (Application) Protein
Product Type Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.