Learn More
Description
- The wide-pore (300 Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
Specifications
Specifications
| Diameter (Metric) | 2.1 mm |
| Length (Metric) | 150 mm |
| Particle Shape | Spherical |
| Particle Size | 1.7 μm |
| Pore Size | 300 Å |
| Quantity | 1/Ea. |
| USP Type | L1 |
| Endcapped | Yes |
| For Use With (Application) | Peptides |
| Product Type | Column |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.