Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corporation ACQUITY UPLC Glycoprotein BEH Amide Column with Glycoprotein Performance Test Standard

Catalog No. 501384494 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
50 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
50-138-4494 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-138-4494 Supplier Waters Corporation Supplier No. 176003700
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Waters Glycoprotein BEH Amide Columns were primarily designed for the analysis of intact glycoproteins, glycoprotein fragments, and glycopeptides.

  • Each batch of Glycoprotein BEH Amide is specifically quality control tested with the Waters Glycoprotein Performance Test Standard to help ensure batch to batch consistency.
  • This kit includes one Glycoprotein BEH Amide column and one vial of Waters Glycoprotein Performance Test Standard.
  • The standard is included to be able to immediately benchmark the column upon receipt and can be reordered as a separate part for continued monitoring.
  • Each batch of digestion standard is prepared under strict quality control criteria confirmed on the included certificate of analysis.

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 50 mm
Particle Shape Spherical
Particle Size 1.7 μm
Pore Size 300 Å
Quantity 1/Ea.
USP Type L68
Endcapped No
For Use With (Application) Glycan
Product Type Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.