Learn More
Waters Corporation ACQUITY UPLC Protein BEH SEC Column and BEH200 SEC Protein Standard Mix
Shop All Waters Chromatography Solutions Products
Description
- Fully optimized for the analysis of proteins and their aggregates with molecular weights ranging from 10,000 to 450,000 Daltons, the total system solution will routinely deliver fast, accurate aggregate determinations up to 10 times quicker that traditional HPLC-based SEC methods.
Specifications
Specifications
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 30 mm |
| Particle Shape | Spherical |
| Particle Size | 1.7 μm |
| Pore Size | 200 Å |
| Quantity | 1/Ea. |
| USP Type | L33 |
| Endcapped | No |
| For Use With (Application) | Protein |
| Product Type | Column |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.