Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corporation ACQUITY UPLC Protein SEC Column and BEH450 SEC Protein Standard Mix

Catalog No. 50465951 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
30 mm
150 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
50-465-951 30 mm
50-465-950 150 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-465-951 Supplier Waters Corporation Supplier No. 176002997
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ACQUITY UPLC BEH450 SEC Column based upon the highly successful BEH particle technology.

  • Based upon the highly successful BEH particle technology, the ACQUITY UPLC BEH450 SEC column is fully optimized for the analysis of proteins and their aggregates with molecular weights ranging from 100,000 to 1,500,000 Daltons, the total system solution will routinely deliver fast, accurate aggregate determinations up to 10 times quicker that traditional HPLC-based SEC methods.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 30 mm
Particle Shape Spherical
Particle Size 2.5 μm
Pore Size 450 Å
Quantity 1/Ea.
USP Type L33
Endcapped No
For Use With (Application) Protein
Product Type Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.