Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACRBP Antibody, Novus Biologicals™
SDP

Catalog No. p-200042554 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB406604 25 μL
NBP185407 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB406604 Supplier Novus Biologicals Supplier No. NBP18540725UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

ACRBP Polyclonal specifically detects ACRBP in Human, Monkey, Rhesus Macaque samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ACRBP
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:5000 - 1:10000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:5000 - 1:10000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q8NEB7
Gene Alias acrosin binding protein, acrosin-binding protein, Cancer/testis antigen 23, Cancer/testis antigen OY-TES-1, CT23FLJ51160, OY-TES-1, proacrosin binding protein sp32, Proacrosin-binding protein sp32, SP32
Gene Symbols ACRBP
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:AKRVLCSQPVSILSPNTLKEIEASAEVSPTTMTSPISPHFTVTERQTFQPWPERLSNNVEELLQSSLSLGGQEQAPEHKQEQGVEHRQEPTQEHKQE
Molecular Weight of Antigen 61 kDa
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84519
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.