Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACRBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558649
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558649 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558649 Supplier Invitrogen™ Supplier No. PA558649
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQDSTQASTP GSPLSPTEYE RFFALLTPTW KAETTCRLRA THGCRNPTLV QLDQYENHGL VPDGAVCSNL PYASWFESFC QFTHYRCS Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

The protein encoded by this gene is similar to proacrosin binding protein sp32 precursor found in mouse, guinea pig, and pig. This protein is located in the sperm acrosome and is thought to function as a binding protein to proacrosin for packaging and condensation of the acrosin zymogen in the acrosomal matrix. This protein is a member of the cancer/testis family of antigens and it is found to be immunogenic. In normal tissues, this mRNA is expressed only in testis, whereas it is detected in a range of different tumor types such as bladder, breast, lung, liver, and colon. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACRBP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACRBP
Gene Accession No. Q8NEB7
Gene Alias ACRBP; acrbp {ECO:0000312; acrosin binding protein; acrosin-binding protein; Cancer/testis antigen 23; cancer/testis antigen OY-TES-1; CT23; EMBL:AAH79212.1}; OY-TES-1; proacrosin binding protein; proacrosin binding protein sp32; proacrosin-binding protein sp32; SP32; testicular tissue protein Li 10
Gene Symbols ACRBP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACRBP. Recombinant protein control fragment (Product #RP-97712).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84519
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.